uvex ProfiErgo ENB 20 Nitrile-Coated Assembly Gloves
Special NBR coating is resistant to oil and grease, while also being touchscreen compatible. The high-quality cotton interlock liner enhances user comfort, feels pleasant against the skin, and helps to regulate glove temperature.
11.85 €
  • Thick, special NBR coating is resistant to oil and grease
  • High-quality cotton interlock liner is comfortable, feels pleasant against the skin, and aids in regulation of the glove climate
  • Enhanced grip capabilities in damp, wet, and oily areas
  • Gloves are suitable for operating touch displays
  • Ideal for a variety of applications including assembly, control work, and maintenance
Product Code Brand Size Price ??product.searchResults.label.quantityAvail??
Quantity
15306942
uvex
6014800
6 11.85 €
Each
Add to basket
15316942
uvex
6014801
7 11.85 €
Each
Add to basket
15326942
uvex
6014802
8 11.85 €
Each
Add to basket
15336942
uvex
6014803
9 11.85 €
Each
Add to basket
15396932
Uvex™
6014804
10 11.85 €