Learn More
RBM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14363-1-AP-150UL
 View more versions of this product
Product Code. 16842974
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This gene is a member of the glycine-rich RNA-binding protein family and encodes a protein with one RNA recognition motif domain. Expression of this gene is induced by cold shock and low oxygen tension. A pseudogene exists on chromosome 1. Multiple alternatively spliced transcript variants that are predicted to encode different isoforms have been characterized although some of these variants fit nonsense-mediated decay criteria.
Specifications
Specifications
RBM3
Polyclonal
Unconjugated
RBM3
IS1 RNPL, Putative RNA binding protein 3, RBM3, RNA binding motif protein 3, RNPL
Rabbit
Antigen Affinity Chromatography
RUO
114488, 19652, 5935
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O89086, P98179, Q925G0
RBM3
RBM3 Fusion Protein Ag5934
150 μL
Primary
Human, Mouse, Rat