Learn More
SCAT Round Canisters
SCAT round canisters are ideal for collecting and disposing of smaller quantities of solvents due to their compact dimensions. Several SCAT round canisters can be easily stationed at the HPLC system and equipped with SCAT SafeyWasteCaps or the SCAT Universal Waste Hub 'Jan'.
Brand: SCAT 107722
77.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
- Available in the common threads S65, S70/71 and B83
- Filling capacities range from very compact 2 L to 4 L, 10 L, 25 L, 50 L and 60 L for the collection of voluminous solvents
Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances
SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.
Specifications
1.3 gal. | |
S65 | |
Round canister 5 L, S65, PE-HD, dimensions (W x H x D): 167 x 330 x 167 mm | |
Reusable | |
HDPE | |
Canister |
5 L | |
White (Translucent) | |
167 x167 x 330 mm | |
with handle and measuring scale | |
1 Ea. | |
Round |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.