Proteintech SLC22A14 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Additional Details:
Weight:
0.00010kg
Specification
Antigen | SLC22A14 |
---|---|
Applications | Western Blot,ELISA |
Applications | Western Blot,ELISA |
Classification | Polyclonal |
Concentration | 750 μg/mL |
Conjugate | Unconjugated |
Gene Symbols | MGC163415; OCTL2; OCTL4; ORCTL4 |
Host Species | Rabbit |
Immunogen | Fusion Protein |
Purification Method | Antigen affinity purification |
Quantity | 20 μL |
Regulatory Status | RUO |
Primary or Secondary | Primary |
Gene ID (Entrez) | 9389 |
Target Species | Human |
Content And Storage | Store at -20°C. Stable for one year after shipment. |
Antibody Molecular Weight | 67 kDa |
Product Type | Polyclonal Antibody |
Form | Liquid |
Isotype | IgG |