JT Baker Sodium Carbonate, Anhydrous, Multi-Compendial, N.F., J.T. Baker™


Overview
739.91 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Code: NEW

Additional Details:
Additional Details: CAS Number: 497-19-8 Weight: 2.50000kg


Quantity 2.5 kg
Packaging Poly Pail

Product Code. 12604735

Quantity Price
1 949.0  € / 2.50kg
Estimated Shipment
31-10-2025
Add to basket


Description and Specification

Specification

Chemical Name or Material Sodium Carbonate
Name Note Anhydrous
CAS 497-19-8
Quantity 2.5 kg
Molecular Formula CNa2O3
MDL Number MFCD00003494
Synonym sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
Synonym sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
Synonym sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
Synonym sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
Synonym sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
Synonym sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
Synonym sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
Synonym sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
Synonym sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
Synonym sodium carbonate,disodium carbonate,soda ash,carbonic acid disodium salt,calcined soda,sodium carbonate, anhydrous,carbonic acid, disodium salt,washing soda,sodiumcarbonate,solvay soda
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES [Na+].[Na+].[O-]C([O-])=O
IUPAC Name disodium;carbonate
Molecular Weight (g/mol) 105.99
PubChem CID 10340
ChEBI CHEBI:29377
Formula Weight 105.99
Grade Multi-Compendial/NF
Packaging Poly Pail
Specific Gravity 1L = 2.53kg
Safety and Handling

[Sodium Carbonate]

[Warning]

[{code=EYE DAM. 2, sequence=1, description=Serious eye damage/eye irritation Category 2}]

[{code=H319, sequence=1, description=Causes serious eye irritation.}]

[{code=P280, sequence=51, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P305+P351+P338, sequence=52, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}]

[{code=MIXTURE LIST, sequence=1, description=Contains : Sodium carbonate}]

[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]


Product Identifier
  • Sodium Carbonate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319- Causes serious eye irritation.

Precautionary Statement
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental Information
  • MIXTURE LIST- Contains : Sodium carbonate