Thermo Scientific Alfa Aesar Sodium chloride, ACS, 99.0% min, Thermo Scientific Chemicals

CAS: 7647-14-5 | ClNa | 58.44 g/mol

Overview
178.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Code: NEW

Additional Details:
Additional Details: CAS Number: 7647-14-5 Weight: 10.00000kg


Quantity 10 kg

Disclaimers: RUO – Research Use Only

Product Code. 11351679

Quantity Price
1 183.0  € / 10kg
In stock, ready to ship: 3
Add to basket


  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
Synonym sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
PubChem CID 5234
ChEBI CHEBI:26710
SMILES [Na+].[Cl-]

Specification

Chemical Name or Material Sodium chloride
Name Note For analysis ACS
CAS 7647-14-5
Physical Form Crystalline
Quantity 10 kg
Assay Percent Range 99.0% min
Molecular Formula ClNa
MDL Number MFCD00003477
Synonym sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
Synonym sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
Synonym sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
Synonym sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
Synonym sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
Synonym sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
Synonym sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
Synonym sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
Synonym sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
Synonym sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES [Na+].[Cl-]
IUPAC Name sodium chloride
Molecular Weight (g/mol) 58.44
PubChem CID 5234
ChEBI CHEBI:26710
Formula Weight 58.44
Density 2.165g/mL
Alternative Products
Documentation

Technical documents