Product Code | Brand | Quantity | Packaging | Price | ??product.searchResults.label.quantityAvail?? |
|
|||
---|---|---|---|---|---|---|---|---|---|
15855188
Safety data sheet (SDS) |
Thermo Scientific
J21618.36
|
500 g | Plastic bottle |
62.4 €
500g |
|
||||
15805338
Safety data sheet (SDS) |
Thermo Scientific
J21618.A1
|
1 kg | Plastic bottle |
86.95 €
1kg |
|
||||
15815338
Safety data sheet (SDS) |
Thermo Scientific
J21618.A7
|
5 kg | Plastic bottle |
158.0 €
5kg |
|
||||
15825338
Safety data sheet (SDS) |
Thermo Scientific
J21618.A9
|
10 kg | Plastic bottle |
207.0 €
10kg |
|
Sodium Chloride, ACS, ≥99%, Ultrapure
Used for precipitation of DNA | CAS: 7647-14-5 | ClNa | 58.44 g/mol
62.4 € - 207.0 €
Chemical Identifiers
CAS | 7647-14-5 |
---|---|
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Disclaimers:
RUO – Research Use Only
Specification
Boiling Point | 1413°C |
---|---|
Chemical Name or Material | Sodium Chloride |
Melting Point | 801°C |
CAS | 7647-14-5 |
Physical Form | Crystalline Powder |
Quantity | 500 g |
Molecular Formula | ClNa |
MDL Number | MFCD00003477 |
Synonym | sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex |
Synonym | sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex |
Synonym | sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex |
Synonym | sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex |
Synonym | sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex |
Synonym | sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex |
Synonym | sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex |
Synonym | sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex |
Synonym | sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex |
Synonym | sodium chloride,salt,table salt,halite,saline,rock salt,common salt,sodium chloride nacl,dendritis,purex |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
SMILES | [Na+].[Cl-] |
IUPAC Name | sodium;chloride |
Molecular Weight (g/mol) | 58.44 |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
Formula Weight | 58.44 |
Percent Purity | ≥99% |
Grade | ACS |
Packaging | Plastic bottle |
Specific Gravity | 2.165 |
Density | 2.165 |