missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Solution, 5% Available Chlorine, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. p-5383214 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
HDPE Pressure Bottom Poly Bottle
Pressure Bottom Poly Bottle
Unit Size:
4 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
16499169 500 mL HDPE Pressure Bottom Poly Bottle 500mL
15538614 4 L Pressure Bottom Poly Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499169 Supplier Avantor J.T.Baker Supplier No. 941601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Quantity 500 mL
Formula Weight 74.44
Percent Purity 5% (Available Chlorine)
Grade BAKER ANALYZED™ Reagent
Packaging HDPE Pressure Bottom Poly Bottle
Specific Gravity 1L = 1.11kg
Chemical Name or Material Sodium Hypochlorite Solution
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301-IF SWALLOWED:.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.