missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Stainles Steel Clamp for Flasks

For use with Grant Instruments UP-168 platform that fits in ES-20/80

Brand:  Grant Instruments™ HSC-500

 View more versions of this product

Product Code. 17735019

  • 86.10 € / Each
Only null left
Estimated Shipment: 30-07-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Clamp, stainless steel,flask, UP-168, platform,ES-20/80, Grant Instruments

TRUSTED_SUSTAINABILITY
Specifications

Specifications

500 mL flask
Clamp
Tight fit clamp 500 mL - 105 mm dia. for UP-168
UP-168
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.