missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Stainless Steel Jugs

Product Code. p-200009504 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
0.5 L
1 L
2 L
3 L
5 L
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17912132 0.5 L Each
17922102 1 L Each
17972102 2 L Each
17942102 3 L Each
17982102 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17912132 Supplier Sampling Systems™ Supplier No. A406500
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These high quality jugs made from 316L stainless steel have a number of GMP features, making them an ideal choice for transferring liquids in laboratory and production areas.

  • GMP design: Crevice-free, mirror polish, full welds
  • Includes a pouring lip for easy emptying
  • Graduation marks for visual convenience

Specifications

Capacity (Metric) 0.5 L
Material Stainless Steel (316L)
Sterility Non-sterile
Graduated Yes
Certifications/Compliance FDA
Style Handled
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.