missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Standard Ground Joint Funnel
Half-round body
Brand: BRAND™ 94910
60.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
50 mm | |
Stem for NS 19/26 | |
Standard Ground Joint Funnel |
Polypropylene | |
10 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction