missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sampling Systems™ Sterile Liquid and Powder Bottles

Store liquids and powders in a sterile environment with Sterile Liquid and Powder Bottles. Made from FDA and EU conforming HDPE/PP, these liquid tight containers are ideal for use in the pharmaceutical, food, and chemical industry. Available from stock in a range of sizes.
Brand: Sampling Systems™ 8404H-250S
This item is not returnable.
View return policy
Description
- A range of high quality, Wide Mouth Sterile Bottles.
- The bottles have a body made from HDPE and a screw on cap made from PP.
- All polymers used for these bottles are made from FDA conforming polymers.
- These sterile bottles are ideal for use in laboratories and production areas.
- The bottles have a wide neck which makes them ideal when working with powders as well as liquids.
- The packaging process takes place in a Class 7 Cleanroom and each bottle is packed into a laminate film bag.
- After packing the bottles are sterilised by gamma irradiation.
- Simply unwrap and use.
- Made from FDA, EC 1935/2004, and EU 10/2011 conforming HDPE/PP.
Specifications
| 250 mL | |
| Individually Bagged | |
| Wide, Threaded |
| HDPE | |
| Sterile | |
| 131 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction