Learn More
Synoptics™ HILED Lighting Option
GREENER_CHOICE
HILED option for illumination of multiplex fluorescence blots - for use in Syngene Gbox
Brand:  Synoptics™ HILED-RGBIR
Product Code. 15709499
  • 8060.00 € / Each
Estimated Shipment: 25-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • HILED Lighting option for GBOX
  • Lighting units: Red, Green, Blue and Infra-Red
  • Includes gantry and electronics
HILED, Multiplex. Fluorescence, westerns
Specifications
Specifications
Syngene
Syngene Gbox
Blue, green, red, infrared
Fluorescence westerns
1Each
HILEDS