Agilent BioTek Take3 Calibration Solution
Take3 Calibration Solution, 25 mL is for use with Microplate Washers and Readers
Additional Details:
Additional Details: Weight: 0.50000kg
Product Code. 16533042
Quantity Price
1 67.1  € / Each
Estimated Shipment
17-10-2025
Add to basket
Description and Specification
Specification
Product Type Solution Concentrate
For Use With (Equipment) Agilent BioTek Microplate Washers and Readers
Capacity (Metric) 25 mL
For Use With (Application) Calibration Solution
  • Concentrated solution for Take3 calibration
  • 1 year shelf life