Thermo Scientific Alfa Aesar Tetradecanonitrile, 98%, Thermo Scientific™


Overview
24.50 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Code: NEW

Additional Details:
Additional Details: CAS Number: 629-63-0 Weight: 0.02500kg


Quantity 25 g

Product Code. 11418597

 N/A
N/A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description and Specification

Chemical Identifiers

CAS 629-63-0
Molecular Formula C14H27N
Molecular Weight (g/mol) 209.38
MDL Number MFCD00019848
InChI Key MLRCLPRHEOPXLL-UHFFFAOYSA-N
Synonym myristonitrile,tetradecanonitrile,1-cyanotridecane,myristyl nitrile,n-tridecyl cyanide,myrystyl nitrile,tridecyl cyanide,c14-18 nitriles,c14-c18 alkylnitrile,acmc-209nb3
PubChem CID 64678
IUPAC Name tetradecanenitrile
SMILES CCCCCCCCCCCCCC#N

Specification

Chemical Name or Material Tetradecanonitrile
CAS 629-63-0
Melting Point ∼19°C
Density 0.83
Boiling Point 226°C to 227°C (100 mmHg)
Odor Odorless
Molecular Formula C14H27N
Refractive Index 1.4415
MDL Number MFCD00019848
Quantity 25 g
Beilstein 1705598
Synonym myristonitrile,tetradecanonitrile,1-cyanotridecane,myristyl nitrile,n-tridecyl cyanide,myrystyl nitrile,tridecyl cyanide,c14-18 nitriles,c14-c18 alkylnitrile,acmc-209nb3
Synonym myristonitrile,tetradecanonitrile,1-cyanotridecane,myristyl nitrile,n-tridecyl cyanide,myrystyl nitrile,tridecyl cyanide,c14-18 nitriles,c14-c18 alkylnitrile,acmc-209nb3
Synonym myristonitrile,tetradecanonitrile,1-cyanotridecane,myristyl nitrile,n-tridecyl cyanide,myrystyl nitrile,tridecyl cyanide,c14-18 nitriles,c14-c18 alkylnitrile,acmc-209nb3
Synonym myristonitrile,tetradecanonitrile,1-cyanotridecane,myristyl nitrile,n-tridecyl cyanide,myrystyl nitrile,tridecyl cyanide,c14-18 nitriles,c14-c18 alkylnitrile,acmc-209nb3
Synonym myristonitrile,tetradecanonitrile,1-cyanotridecane,myristyl nitrile,n-tridecyl cyanide,myrystyl nitrile,tridecyl cyanide,c14-18 nitriles,c14-c18 alkylnitrile,acmc-209nb3
Synonym myristonitrile,tetradecanonitrile,1-cyanotridecane,myristyl nitrile,n-tridecyl cyanide,myrystyl nitrile,tridecyl cyanide,c14-18 nitriles,c14-c18 alkylnitrile,acmc-209nb3
Synonym myristonitrile,tetradecanonitrile,1-cyanotridecane,myristyl nitrile,n-tridecyl cyanide,myrystyl nitrile,tridecyl cyanide,c14-18 nitriles,c14-c18 alkylnitrile,acmc-209nb3
Synonym myristonitrile,tetradecanonitrile,1-cyanotridecane,myristyl nitrile,n-tridecyl cyanide,myrystyl nitrile,tridecyl cyanide,c14-18 nitriles,c14-c18 alkylnitrile,acmc-209nb3
Synonym myristonitrile,tetradecanonitrile,1-cyanotridecane,myristyl nitrile,n-tridecyl cyanide,myrystyl nitrile,tridecyl cyanide,c14-18 nitriles,c14-c18 alkylnitrile,acmc-209nb3
Synonym myristonitrile,tetradecanonitrile,1-cyanotridecane,myristyl nitrile,n-tridecyl cyanide,myrystyl nitrile,tridecyl cyanide,c14-18 nitriles,c14-c18 alkylnitrile,acmc-209nb3
InChI Key MLRCLPRHEOPXLL-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCC#N
IUPAC Name tetradecanenitrile
Molecular Weight (g/mol) 209.38
PubChem CID 64678
Formula Weight 209.38
Percent Purity 98%
Documentation

Technical documents