Brand™ PMMA Semi-Micro Cuvettes
These macro or semi-micro cuvettes that are available in PS and PMMA are ideal for kinetic measurements.
Overview
15.37 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Code: A0
Additional Details:
Additional Details: Weight: 0.27000kg
Product Code. 10038810
Quantity Price
1 36.4  € / Pack of 100
In stock, ready to ship: 215
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Type Semi-micro Cuvette
Capacity (Metric) 1.5 to 3 mL
Material PMMA
Quantity 100Pack
  • Macro or semi-micro cuvettes available in PS and PMMA
  • Ideal for kinetic measements
  • Windows are recessed to protect from scratches, arrow mark indicates optical path orientation
  • Plastibrand cuvettes are delivered grouped by mold cavity number to ensure lowest variation of extinction coefficient
  • Useable wavelength > 300 nm
  • Standard deviation at 320 nm: ±0.004 extinction units