412.53 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Max. Temperature | 320/340°C |
---|---|
Diameter (Metric) | 0.32 mm |
Diameter (Metric) Inner | 0.32 mm |
Product Line | TRACE |
Stationary Phase | Trace™ TR-5 |
USP Type | G27, G36 |
For Use With (Application) | Free Fatty Acids Analyis,GC/MS for Free Fatty Acids,GC/MS for Aromatics,GC/MS for Flavors,GC/MS for Low Polarity Pesticides |
For Use With (Application) | Free Fatty Acids Analyis,GC/MS for Free Fatty Acids,GC/MS for Aromatics,GC/MS for Flavors,GC/MS for Low Polarity Pesticides |
For Use With (Application) | Free Fatty Acids Analyis,GC/MS for Free Fatty Acids,GC/MS for Aromatics,GC/MS for Flavors,GC/MS for Low Polarity Pesticides |
For Use With (Application) | Free Fatty Acids Analyis,GC/MS for Free Fatty Acids,GC/MS for Aromatics,GC/MS for Flavors,GC/MS for Low Polarity Pesticides |
For Use With (Application) | Free Fatty Acids Analyis,GC/MS for Free Fatty Acids,GC/MS for Aromatics,GC/MS for Flavors,GC/MS for Low Polarity Pesticides |
Film Thickness | 0.25 μm |
Length (Metric) | 15 m |
Polarity | Non-Polar |
Description | 15 m x 0.32 mm x 0.25 μm |