304.91 EUR valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Use promo code "24124" to get your promotional price.
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
---|---|
Lid | With Lid |
Culture Area | 0.084 cm2/well |
Array | 16 x 24 |
Packaging | 10 Pk., 30 Cases |
Product Line | Nunc |
Color | White |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 120 μL |
No. of Wells | 384 |
Volume (Metric) Working | 10 to 100 μL |
Sterility | Sterile |
Surface Treatment | Cell Culture |
Well Shape | Flat Bottom (F-well) |
No. per Case | 30 |
No. per Pack | 10 |