Code:
99
Eppendorf™ Swing Bucket Rotor for Benchtop Centrifuge
Use these rotors for a wide range of molecular applications. Eppendorf™ Swing Bucket Rotors are designed for use with Eppendorf benchtop centrifuges.
Additional Details:
Weight:
2.23560kg
Specification
Type | Swinging Bucket Rotor |
---|---|
Max. RCF | 3,000 x g |
Max. Speed | 4,400 rpm |
For Use With (Equipment) | A-4-38 |
Includes | 4 x 85mL buckets |
No. of Places | 4 |