Learn More
Fisherbrand™ ProFill HPLC Syringe Filters, Regenerated Cellulose Membrane
HPLC performance tested
Brand:  Fisherbrand™ 30 16 2089
 View more versions of this product
Product Code. 10270225
  • 298.00 € / Pack of 100
Only null left
In Stock
Add to basket
Description
Description
  • ProFill
  • HPLC Syringe Filter
  • Regenerated Cellulose (RC)
  • Low extractable membranes and housing
  • Polypropylene housing
  • Enhanced Luer Lock inlet which prevents leakage
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Brown
0.45 μm
<0.137 mL
No
100/Pk.
30 mm
Regenerated Cellulose
4.91 cm2
PE-Bag, additionally packed in a PP-Box
6.2bar
HPLC Syringe Filter