Code:
69
Cytiva Whatman™ Qualitative Filter Paper: Grade 595 ½ Pleated Circles
Pleated to save time and achieve optimal fit in conical filter funnels. Whatman™ Qualitative Filter Paper: Grade 595 ½ Pleated Circles provide extra surface area to increase total loading capacity.
Additional Details:
Weight:
0.42000kg
Specification
Description | Grade 595 ½ Qualitative Filter Paper Folded (Prepleated), 270 mm |
---|---|
Format | Pre-pleated Circle |
Material | Cellulose |
Particle Retention | 7 μm |
Pore Size | 7 μm |
Filtration Speed | Approx. 80 s |
Flow Rate | Medium fast |
For Use With (Application) | A prepleated filter used for many routine analytical applications in different industries (e.g., particle separation from food extracts or filtration of solids from digested environmental samples, e.g., for ICP/AAS analysis). |
Grade | Grade 595 ½ |
Quantity | 100 Pk. |
Thickness (Metric) | 150 μm |
Diameter (Metric) | 270 mm |