Learn More
BRAND™ Microcentrifuge Tubes Without Screw Caps
Manufactured from polypropylene and complies with DIN EN 285 standards. BRAND™ Microcentrifuge Tubes Without Screw Caps are graduated with frosted marking area that allows for sample identification or other notations. Colored screw caps and cap inserts available for separate purchase.
Brand:  BRAND™ 780734
111.15 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10354371
  • 146.00 € / Pack of 1000
Estimated Shipment: 04-06-2025
Explore more special offers
Add to basket
Description
Description
  • Manufactured from polypropylene and complies with DIN EN 285 standards.
  • Graduated.
  • Frosted marking area allows for sample identification or other notations.
  • Operating range: -196°C to 121°C.
  • Autoclavable at 121°C (2 bar)
  • Colored screw caps and cap inserts available for separate purchase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIN EN 285
2 mL
Conical bottom
1000
Non-sterile
13 mm
Autoclavable
Polypropylene
44 mm
–196 to 121°C
20,000 G