Learn More
Thermo Scientific™ 384 Well White Plate, Non-Treated Surface, Non-Sterile, Pack of 25
Color:White; Sterility:Non-sterile; Lid:Without Lid
Brand: Thermo Scientific™ 262360
780.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Improve background, autofluorescence and crosstalk with Thermo Scientific™ Nunc 384-Well Polystyrene Black and White Microplates.
- Black microplates are recommended for fluorescence measurements; with minimum back-scattered light and background fluorescence
- White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
0.05 cm2 | |
25/Pk., 100/Cs. | |
White | |
Polystyrene | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
25 |
Without Lid | |
16 x 24 | |
Nunc | |
Microplate | |
120 μL | |
10 to 100 μL | |
Non-treated | |
100 | |
100/Cs. |