182.40 EUR valid until 2025-12-31
Use promo code "24367" to get your promotional price.
Use promo code "24367" to get your promotional price.
Color | Clear |
---|---|
Capacity (Metric) | 0.5 mL |
Material | Polypropylene |
Quantity | 1000 Tubes |
Content And Storage | Store at room temperature. |
Product Line | Invitrogen |
Shipping Condition | Room Temperature |
Dnase Free | RNase-Free, DNase-Free |
Autoclavable | Autoclavable |
Closure Type | Snap Cap |
Closure Included | Yes |
Graduated | Yes |
Product Type | Microfuge Tube (RNase-free) |
Surface Type | Graduated |