Learn More
Platinum™, SYBR™ Platinum™ SYBR™ Green qPCR SuperMix-UDG
Provides a sensitive and convenient system for real-time PCR detection with SYBR™ Green I dye.
Brand:  Platinum™, SYBR™ 11733046
 View more versions of this product
Product Code. 10633863
  • 1720.00 € / 500 reactions
Includes: Platinum SYBR Green qPCR SuperMix-UDG (2 × 1.25mL), 50mM Magnesium Chloride (1mL), ROX Reference Dye (100μL), 20X BSA (300μL)
Description
Description
The SuperMix format combines amplification and detection reagents into a ready-to-go mix to make set-up easy and ensure reproducible results.
  • Platinum™ Taq DNA Polymerase for sensitive amplification from as few as 10 copies of target
  • 100% dUTP and UDG enzyme for superior control of carryover contamination, saving time by reducing the number of failed experiments
  • Separate tubes of ROX reference dye and BSA for easy optimization on 96-well plates and glass capillary tube instruments
  • SYBR™ Green I dye for simple and easy detection of real-time product
Order Info
Shipping Conditions: Dry Ice
TRUSTED_SUSTAINABILITY
Specifications
Specifications
• Platinum™ SYBR™ Green qPCR SuperMix-UDG (12.5 mL)
• 50 mM Magnesium Chloride (2 × 1 mL)
• ROX Reference Dye (500 μL)
• 20X BSA (1.3 mL)
Sufficient reagents are provided for 500 reactions of 50 μL each. Stored at -20°C.
Tube
qPCR
Standard
Platinum™, SYBR™
500 reactions
500 Reactions of 50 μL Each
500 Reactions
12.5 mL
SYBR
High
Taq DNA Polymerase
7000 System, 7300 System, 7500 System, 7700 System, 7900HT System, GeneAmp 5700, StepOne™, StepOnePlus™, ViiA™ 7 System, Cepheid SmartCycler, BioRad iCycler iQ
Real Time PCR SYBR Master Mix
Dry Ice
Gene Expression
DNA (Genomic), cDNA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.