Cytiva L-Glutamine
Supplement serum- and protein-free media using this amino acid.
Overview
Code: A0
Additional Details:
Additional Details: Weight: 0.50000kg
Product Code. 10691233
Quantity Price
1 37.2  € / 500mL
Estimated Shipment
15-07-2025
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Safety and Handling
Description and Specification
Specification
Content And Storage Store frozen
Description L-Glutamine, 200mM Solution (29.2mg/mL-Glutamine in 0.85% NaCl), 500mL
Concentration 29.2 mg/mL
Form Liquid
Sterilization Method 0.1 μm Sterile-filtered
Quantity 500 mL
  • Format: Liquid
Alternative Products
Documentation Safety and Handling
[{code=EUH210, sequence=91, description=Safety data sheet available on request.}]
[{code=UNKNOWN AQUATIC, sequence=1, description=Contains 3% of components with unknown hazards to the aquatic environment.}]
Hazard Statement
  • EUH210- Safety data sheet available on request.
Supplemental Information
  • UNKNOWN AQUATIC- Contains 3% of components with unknown hazards to the aquatic environment.