Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tube
Store peptide, antibody or virus samples in these LoBind microcentrifuge tubes. Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tubes are ideal for work in protein research, mass spectrometry and protein array sample preparation.
Overview
12.19 EUR valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Code: 96
Additional Details:
Additional Details: Weight: 0.18000kg
Product Code. 10708704
Quantity Price
1 13.35  € / Pack of 100
In stock, ready to ship: 638
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Specification
Closure Material Polypropylene
Capacity (Metric) 1.5 mL
Color Transparent
Material Polypropylene
For Use With (Application) Protein LoBind consumables are best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus samples
No. per Pack 100 Tubes (2 bags of 50)
Volume (Metric) 1.5mL
Sterility PCR Clean
Max. RCF 18,000 x g
  • Problem free handling with the tiniest volumes/concentrations of a variety of proteins
  • Preliminary handling of tubes by saturation with BSA or siliconisation is not required in order to guarantee the recovery of the proteins in the plastic tube
  • Recovery rates of more than 99% are achievable
  • Enzyme activities, for example, are not reduced by this tube, and denaturing effects caused by interaction with the surface are almost completely excluded
  • No undesired side effects of a surface coating
  • Tubes are available in Eppendorf LoBind™ material for maximum recovery of valuable samples
  • Maximum protein recovery (over 97%, BSA 1μg/mL)
  • Free from surface coating, e.g., silicone
  • Free from DNA, DNase, RNase and ATP (PCR* clean)
  • Special polypropylene material manufactured under optimized processing conditions
  • High degree of centrifugation durability, rated to 18, 000xg
  • High degree of transparency
  • Safe-Lock lid