Learn More
Thermo Scientific™ Nalgene™ Utility Funnels
GREENER_CHOICE
Replace glass lab funnels with these utility funnels, which are an excellent shatter-proof alternative.
Brand:  Thermo Scientific™ 4256-0414
44.25 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10709041
  • 44.70 € / Pack of 12
Estimated Shipment: 24-07-2025
Add to basket
Description
Description
Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are shatterproof making them a valuble asset to lab safety programs.
  • 4256-0234 and 4256-0414 are made from shatter-proof, autoclavable polypropylene.
  • 4256-0638 has a large throat opening for faster transfer of materials and is made from shatter-proof high-density polyethylene (Not autoclavable).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
98 mm
18 mm
117 mm
Funnel Utility PP 108mm
12/Pk.; 144/Cs.
270 mL
Polypropylene
Autoclavable
37 mm