Learn More
Falcon™ 50 mL High Clarity Conical Centrifuge Tubes
BIOPHARMA
Product Code. 10788561 Shop All Corning Products
Click to view available options
Packaging:
Bag
Rack
Max. RCF:
16,000 x g
Unit Size:
Pack of 500
Includes
Blue 33mm polyethylene flat-top screw caps (threaded to fit Corning Falcon tubes only).
Tested to withstand centrifugation of 16,000 RCF in a fully supported rotor with room temperature water. Falcon 50mL Conical Centrifuge Tubes are ideal for cell centrifugation; pelleting; separation by density gradients.
Suitable for molecular biology applications including concentrating bacteria for DNA isolation, purification and precipitation of nucleic acids; sample storage (ambient temperature to -80°C); and centrifugation of precipitates. 
  • Hydrophobic, biologically inert surface for good cell or protein recovery
  • Dark blue graduations in 5mL increments
  • White writing area resists solvents
  • Irradiated for sterility
 Clarified Polypropylene
  • Resistant to alcohols and mild organic solvents (not recommended for extraction procedures)
  • Translucent wall for easy viewing
  • Stable from -80°C to +121°C (-112°F to +250°F)
  • Withstand centrifugal forces to 16,000 x g RCF
Order Info
Available in bags or recyclable polystyrene racks of 25, cases of 500.
Compliance
Manufactured under ISO 9001: 2000 standard and FDA Quality System Regulation in Chapter 1, Title 21, Section 820 of CFR.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001:2000, FDA, CFR
Closure Color Blue
Diameter (English) Outside 1.2 in.
Color Clear
Capacity (Metric) 50 mL
Material Polypropylene
For Use With For DNA isolation, purification and precipitation of nucleic Acids, cell centrifugation applications
No. per Case 500
Packaging Bag
Sterility Sterile
Shape Conical
Length (Metric) 115 mm
Capacity (English) 1.69 oz.
Bottom Shape Conical
Writing Area White Marking Spot
Diameter (Metric) 30 mm
Product Type Centrifuge Tube
Max. RCF 16,000 x g