Fisherbrand™ Soda Lime Glass Funnel
Plain glass funnel manufactured from soda lime glass. Fisherbrand™ Soda Lime Glass Funnel is ideal for use with funneling liquids in laboratory applications.
Overview
Code: 41
Additional Details:
Additional Details: Weight: 0.03780kg
Product Code. 11522423
Quantity Price
1 2.2  € / Each
In stock, ready to ship: 4
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification
Specification
Diameter (Metric)Top 50mm
Material Soda Lime Glass
Length (Metric) Stem 50mm
Plain glass funnel manufactured from soda lime glass.