Learn More
Fisherbrand™ 24mm PP Screw Seal, White, Center Hole, 24-400 Thread, Assembled Septum
333.31 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
- No time consuming and tricky assembly
- No contamination of the liner with sweat/fat normally caused by manual assembly
- Broad variety of different septa materials for almost all applications
Specifications
PTFE/Butyl | |
24 mm | |
Storage | |
2.5 mm |
Gray, Red, White | |
Polypropylene | |
100 | |
Screw Cap |