Use promo code "24112" to get your promotional price.
Gibco™ Advanced DMEM/F-12
Advanced DMEM/F-12 (Dulbecco's Modified Eagle Medium/Ham's F-12) is a widely used basal medium that allows the culture of mammalian cells with reduced Fetal Bovine Serum (FBS) supplementation.
26.80 EUR valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Additional Details:
Weight:
0.59000kg
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specification
Cell Line | MRC-5, SP2, Vero, WI-38 and Jurkat |
---|---|
Classification | Animal Origin |
Concentration | 1 X |
Form | Liquid |
Product Type | DMEM (Dulbecco's Modified Eagle Medium)/F-12 |
Serum Level | Reduced Serum |
Sterility | Sterile-filtered |
Sterilization Method | Sterile-filtered |
With Additives | High Glucose,Phenol Red,Sodium Pyruvate,Non Essential Amino Acids (NEAA) |
With Additives | High Glucose,Phenol Red,Sodium Pyruvate,Non Essential Amino Acids (NEAA) |
With Additives | High Glucose,Phenol Red,Sodium Pyruvate,Non Essential Amino Acids (NEAA) |
With Additives | High Glucose,Phenol Red,Sodium Pyruvate,Non Essential Amino Acids (NEAA) |
Without Additives | No Glutamine,No HEPES |
Without Additives | No Glutamine,No HEPES |
Manufacturing Quality | cGMP-compliant under the ISO 13485 standard |
Product Line | Gibco™ |
Quantity | 500 mL |
Shipping Condition | Room Temperature |
Content And Storage |
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
[Methyl benzoate]
[Warning]
[{code=ACUTE TOX. 4, sequence=1, description=Acute toxicity Category 4}]
[{code=H302, sequence=1, description=Harmful if swallowed.}]
[{code=P301+P312, sequence=51, description=IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Methyl benzoate}]
[{code=GHS07, sequence=1, description=EXCLAMATION MARK}]
Product Identifier
- Methyl benzoate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302- Harmful if swallowed.
Precautionary Statement
- P301+P312- IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental Information
- MIXTURE LIST- Contains : Methyl benzoate