Ansell TouchNTuff™ Disposable Powdered Green Nitrile Gloves
Resists a greater variety of industrial chemicals for longer periods than any other nitrile disposable gloves. Offers up to four times the puncture resistance of comparable natural-latex gloves, and three times the resistance of similar neoprene gloves.
Overview
21.10 EUR valid until 2025-08-31
Use promo code "24157" to get your promotional price.
Code: S3
Additional Details:
Additional Details: Weight: 0.77220kg
Product Code. 11706584
Quantity Price
1 30.45  € / Pack of 100
In stock, ready to ship: 13
Add to basket
  • Description and Specification
  • Alternative Products
  • Documentation
  • Special Offers
Description and Specification
Specification
Certifications/Compliance FDA food handling requirements (21 CFR 177.2600)
Color Teal
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powdered
Product Type Nitrile Gloves
Size X-Large
Thickness (Metric) 0.12 mm
Glove Cuff Style Beaded/Rolled
Quantity 100 Pack
Series 92-500
Length (Metric) 240 mm
  • Contains no natural rubber proteins, which means no risk of Type I allergies for the wearer.
  • Primary skin irritation studies and Insult Patch tests have also shown no evidence of risk from irritation or allergic contact dermatitis.
  • Unique Thin Nitrile Technology formulation offers easy donning and strong grip in wet or dry conditions.
  • Highly versatile and suitable for many different uses.
  • Provides three times the puncture resistance of natural latex and vinyl gloves
  • Exceptionally soft feel
  • Rolled cuff, 0.12mm thickness at the palm
Alternative Products
Documentation Special Offers