Use promo code "25339" to get your promotional price.
Sartorius Microsart™ @Filter 250 in Bags
Use promo code "25339" to get your promotional price.
Specification
Color | White |
---|---|
Disposable | Single-use |
Format | Disc |
Material | Cellulose Nitrate |
Material (Membrane) | Cellulose Nitrate |
Pore Size | 0.2 μm |
Sample Volume | 250 mL |
Sterility | Sterile |
Grid Color | Black |
Type | Filter Tip |
Diameter (Metric) | 47 mm |
[X48 MS @FILTER 250 CN B/N 0.2 µM]
[Danger]
[{code=EYE DAM. 2, sequence=1, description=Serious eye damage/eye irritation Category 2}, {code=FLAM. SOL. 2, sequence=2, description=Flammable solid Category 2}, {code=SKIN IRRIT. 2, sequence=3, description=Skin corrosion/irritation Category 2}]
[{code=H228, sequence=1, description=Flammable solid.}, {code=H315, sequence=2, description=Causes skin irritation.}, {code=H319, sequence=3, description=Causes serious eye irritation.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P241, sequence=52, description=Use explosion-proof electrical/ventilating/lighting/ equipment.}, {code=P260, sequence=53, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=54, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P303+P361+P353, sequence=55, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P305+P351+P338, sequence=56, description=IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.}, {code=P370+P378, sequence=57, description=In case of fire: Use dry sand, water spray, CO2 or alcohol resistant foam to extinguish.}, {code=P405, sequence=58, description=Store locked up.}, {code=P501b, sequence=59, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : nitrocellulose}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
- X48 MS @FILTER 250 CN B/N 0.2 µM
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H228- Flammable solid.
- H315- Causes skin irritation.
- H319- Causes serious eye irritation.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241- Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378- In case of fire: Use dry sand, water spray, CO2 or alcohol resistant foam to extinguish.
- P405- Store locked up.
- P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
- MIXTURE LIST- Contains : nitrocellulose