Learn More
Wheaton™ Borosilicate Glass Base
The Borosilicate Glass Base is a replacement component part, a support base, for the Wheaton™ 90 mm Filtration Assembly. Manufactured from Wheaton™ 33 low extractable borosilicate glass conforming to USP Type I and ASTM E 438 Type I, Class A requirements.
Brand:  Wheaton™ 419455
Product Code. 11718552
  • 304.00 € / Each
Only null left
Estimated Shipment: 08-08-2025
Add to basket
Description
Description
  • Replacement part for 90 mm filtration assembly
  • Wheaton™ 33 low extractable borosilicate glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Wheaton™ 90 mm Filtration Assembly
Borosilicate Glass
1 L Funnel with Screw Collar, Glass Support Base, Filter Support, No. 8 Silicone Stopper
1 Ea.