Learn More
Bel Art™ Polypropylene Sterilizing Trays
Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.
Brand:  Bel Art™ H16264-0000
60.85 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11762555
  • 69.05 € / Each
Only null left
Estimated Shipment: 09-09-2025
Add to basket
Description
Description
Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.
  • Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks
  • Trays are steam autoclavable at 121°C (250°F)
  • Covers are sold separately
Order Info
Covers are sold separately.
Specifications
Specifications
18 L
White
53.3 cm
Sterilizing Tray
Fits 18L Tray Cover, Mfr. No. H162590000
5 in.
15.5 in.
20.3 x 15.5 x 5 in. (53.3 x 39.4 x 12.7 cm)
12.7 cm
Polypropylene
39.4 cm
21 in.
Rectangular
Autoclavable