Learn More
Cytiva Sera-Mag SpeedBeads™ Carboxyl Magnetic Beads, hydrophobic
Excellent for coupling oligos or proteins directly to the beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
Brand:  Cytiva 65152105050350
2077.30 EUR valid until 2025-09-30
Use promo code "24338" to get your promotional price.
 View more versions of this product
Product Code. 11829912
  • 2342.00 € / Each
Description
  • Double shell magnetite beads
  • . High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophobic bead based on crosslinking requirements for the encapsulation layer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5% solids (50mg/mL)
DNA Sample Prep, Protein Purification
1 μm
2°C to 8°C
100 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications