102.04 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Use promo code "25721" to get your promotional price.
Capacity (Metric) | 15 mL |
---|---|
Material | Polypropylene |
No. per Case | 500 |
No. per Pack | 25 |
Packaging | Bulk, easy-open sleeve |
Shape | Conical |
Bottom Shape | Conical |
Closure Type | Plug |
Max. RCF | 8000 x g |