Learn More
Caframo™ Safety Stand for Overhead Stirrers
GREENER_CHOICE
Wide base and H configuration Safety Stand gives superior stability when agitating viscous materials. For use with Caframo™ High Torque and Light Torque Overhead Stirrers.
Brand:  Caframo™ A110BASE
 View more versions of this product
Product Code. 12997096
  • 280.00 € / Each
Only null left
Estimated Shipment: 06-08-2025
Add to basket
Description
Description
  • Safety Stand for High Torque and Light Torque Overhead Stirrers
  • Cast zinc-aluminum alloy coated H base with chemical resistant epoxy paint
  • Includes allen key
  • Two level adjustment knobs
Specifications
Specifications
430 x 416 mm (16.4 x 16.9 in.)
Stand Rod Not Included
1.6 kg
Cast Zinc, Aluminum
Overhead Stirrer
Safety Lab Stand Base
Chemical-resistant Epoxy Paint
3.5 lb.