Learn More
Sartorius™ Cotton Gloves for Weights
Product Code. 13119250 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
1 Pair/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13119250 1 Pair/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13119250 Supplier Sartorius™ Supplier No. YAW21
Only null left
Add to basket
Edge
Add to basket
Prevent damage to the surface of calibration weights. Sartorius™ Cotton Gloves for Calibration Weights allow you to safely and properly handle finely machined calibration weights and maintain balance accuracy.
  • Cotton construction
  • Ensure accuracy of weight and mass standards
  • Eliminates fingerprints and resists contamination by dirt, moisture, and corrosion
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Calibration Weights and Standards
Color White
Material Cotton
Quantity 1 Pair/Pk.