Learn More
Bel-Art™ Roxy M™ Universal Fit Repeating Pipette Tips, Nonsterile
Provide reliable, and precise repeated dispensing from 1 to 5000μL
Brand: Bel-Art™ F37300-0001
This item is not returnable.
View return policy
Description
- Fit most manual repeating pipettors including Roxy M, Eppendorf™ Multipette™ 4780, Brand HandyStep™ S, Fisherbrand™ Repeating Pipetter, Gilson Distriman™, and Scilogex™ StepMate Repeater Pipettor
- Fits many electronic pipetters (digital display will not be enabled)
- Available in 8 sizes with capacities from 0.05 to 50.0 mL to repeatedly dispense volumes from 1 to 5000μL
- Manufactured in an ISO 9001:2000 facility and made of virgin polypropylene with a polyethylene inner plunger
- Adapter required to use 25 mL and 50 mL tips (Mfr. No. F37300-0050 supplied with repeating pipetter and available separately in a 3-pack)
- Each pack of tips is lot-stamped, comes with a Certificate of Conformity, and includes a convenient dosing chart
Specifications
0.5 mL | |
Non-sterile | |
Autoclavable | |
DNase, RNase, ATP, and Pyrogen Free | |
Bagged | |
Repeating Pipettor Tips | |
Eppendorf Multipette 4780, Fisherbrand Repeating Pipette, Gilson Distriman, Handystep S, Roxy M, Gilson Distriman | |
SP Scienceware Roxy M |
Non-filtered | |
Natural | |
ISO 9001:2000 | |
Roxy M, Eppendorf Multipette 4780, Brand HandyStep S, Fisherbrand Repeating Pipetter, VWR Repeating Pipettor, Gilson Distriman, and Scilogex StepMate Repeater Pipettor | |
Standard | |
Single-use | |
Yes | |
Polypropylene, Polyethylene |