173.99 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Use promo code "24135" to get your promotional price.
Disposable | Single-use |
---|---|
Filter Type | Syringe Filter |
For Use With (Equipment) | Syringe |
Material | Polypropylene |
Material (Membrane) | PES |
Pore Size | 0.22 μm |
Sample Volume | 0.5 to 15 mL |
Sterility | Sterile |
Filtration Area | 1.7 cm2 |
For Use With (Application) | Sterile Filtration, Clarification |
Hold up Volume (After Air Purge) | 30 to 100 μL before bubble point |
Housing Material | Polypropylene (PP) |
Inlet Style | Female Luer Lock |
Outlet Style | Male Luer Slip |
Quantity | 50 Pk. |
Type | Syringe Filter |
Volume (Metric) | 15 mL |