Learn More
Thermo Scientific™ Point of Use Filter Inserts
Protect installations and systems from undissolved substances in water with these pre-filters, final filters and sterile filters.
Brand:  Thermo Scientific™ 50155509
165.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15326945
  • 165.83 € / Each
Only null left
Estimated Shipment: 09-10-2025
Add to basket
Description
Description
Protect installations and systems from undissolved substances in water with these pre-filters, final filters and sterile filters. Thermo Scientific™ Point of Use Filter Inserts include 10- and 20-inch pre-filters, final filters and sterile filters for Point of Use systems.
Pre-filters 1μm, 5μm
  • Protect installations and systems from undissolved substances in water
Final filters
  • Protect systems from undissolved substances in water
  • Usually used for final filtration
Sterile filters
  • For final filtration and delivery of organic-free water
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 in.
Prefilter Insert
5 μm
Pre-filter Housings, 20 in. (06.5320, 06.5321, 06.5322)
3200 L/hr. (at D p = 0.2 bar)