Learn More
Thermo Scientific™ Fiberboard Storage Boxes (Colored)
Shop All Thermo Scientific Products
Click to view available options
Color:
Assorted
Black
Blue
Green
Pink
Red
Yellow
Unit Size:
Pack of 12
Description
Store a variety of vials and tubes with Thermo Scientific™, water-repellant, fiberboard storage boxes, for use in ultra-low temperature or cryogenic freezers. Available in packs of 12; choose from yellow, pink, red, blue, green or multi-colored. Grid dividers sold separately.
- Sturdy, water-repellant fiberboard boxes with covers for sample storage
- Available in 2 in. high (5cm)
Ordering Information:
Available in packs of 12: choose from yellow, pink, red, blue, green or multi-colored. Grid dividers sold separately.
Specifications
Specifications
| Color | Blue |
| Material | Fiberboard, Water-repellant |
| Shape | Square |
| Type | Cryo Storage Box |
| For Use With (Equipment) | Ultra-Low Temperature Freezer |
| Quantity | 12/Pk., 12/Cs. |
| Dimensions (L x W x H) | 5 x 5 x 2 in. (12.7 x 12.7 x 5 cm) |
| Length (English) | 5 in. |
| Length (Metric) | 12.7 cm |
| Width (English) | 5 in. |
| Show More |