Honeywell Fluka HYDRANAL™ - Composite 5, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™
Specification
Chemical Name or Material | HYDRANAL™-Composite 5 |
---|---|
Density | 1.15g/mL |
Flash Point | 90°C (194°F) |
Quantity | 6 x 500 mL |
CAS | 111-90-0 |
CAS | 7553-56-2 |
CAS | 288-32-4 |
CAS | 7446-09-5 |
CAS | 68007-08-9 |
CAS | 693-98-1 |
Technical documents
[HYDRANAL-Composite 5]
[Danger]
[{code=CARC. 2, sequence=1, description=Carcinogenicity Category 2}, {code=REPR. 1B, sequence=2, description=Reproductive toxicity Category 1B}, {code=STOT RE 2, sequence=3, description=Specific target organ toxicity after repeated exposure Category 2}]
[{code=H351, sequence=1, description=Suspected of causing cancer.}, {code=H360D, sequence=2, description=May damage the unborn child.}, {code=H373, sequence=3, description=May cause damage to organs.}]
[{code=P260, sequence=51, description=Do not breathe dust/fume/gas/mist/vapours/spray.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}, {code=P284, sequence=53, description=¢In case of inadequate ventilation! wear respiratory protection.}, {code=P308+P313, sequence=54, description=IF exposed or concerned: Get medical advice/attention.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : sulphur dioxide, 1H-Imidazole monohydriodide, iodine, imidazole, 2-Methylimidazole}]
[{code=GHS08, sequence=1, description=HEALTH HAZARD}]
Product Identifier
- HYDRANAL-Composite 5
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H351- Suspected of causing cancer.
- H360D- May damage the unborn child.
- H373- May cause damage to organs.
Precautionary Statement
- P260- Do not breathe dust/fume/gas/mist/vapours/spray.
- P280- Wear protective gloves/protective clothing/eye protection/face protection.
- P284- ¢In case of inadequate ventilation! wear respiratory protection.
- P308+P313- IF exposed or concerned: Get medical advice/attention.
Supplemental Information
- MIXTURE LIST- Contains : sulphur dioxide, 1H-Imidazole monohydriodide, iodine, imidazole, 2-Methylimidazole
-
Science Is Hard Enough Your Bioreagents Shouldn’t Be
Reliable performance. Special prices.Happier researchers.
Buy Now