Learn More
Thermo Scientific™ Barnstead™ Smart2Pure™ Pro Water Purification System
All in One ! The Smart2Pure™ Pro system is a compact system which converts tap water to into ASTM Type 1. The Smart2Pure Pro UV/UF 16 LPH offers the choice of a 30 L or 60 L reservoir for customizable storage capacity and ability to dispense both ASTM Type 1 and Type II water.
Brand:  Thermo Scientific™ 50157873
8238.92 EUR valid until 2025-08-17
Use promo code "24157" to get your promotional price.
 View more versions of this product
Product Code. 15980206
  • 13869.00 € / 1 unit
Focus on Agri-Science Testing
Use promo code 24136 at checkout
View Available Promotions
Only null left
Estimated Shipment: 03-11-2025
Add to basket
Description
Description
All in One ! The Smart2Pure™ Pro system is a compact system which converts tap water to into ASTM Type 1. The Smart2Pure Pro UV/UF 16 LPH offers the choice of a 30 L or 60 L reservoir for customizable storage capacity and ability to dispense both ASTM Type 1 and Type II water.
Lasting economy:
  • Water purification progresses through independent cartridges with Aquastop quick connects for fast replacement.
  • Module 1 reverse osmosis catridges to remove up to 99% of tap water impurities
  • Module 2 includes a polishing cartridge that contains high-quality ultrapure resin for consistent purity and long cartridge life.
Easy-to-operate:
  • Dispensing is easy and features variable speed to control flow.
  • The display can be tilted for optimal reading.
  • Simplified User Interface
Placement Options:
  • Position on laboratory bench
  • Mount it on the wall
New Features:
  • Automatic Standby mode when system is not in use recirculates water through system to maintain purity
  • Leak detector shuts off water supply and system if leak is detected
  • Alerts show on display when to replace consumables
  • Reverse osmosis water quality monitoring
  • Inline ultrafilter with large surface area to remove endotoxins and nucleases.
  • System parameters are code-protected to prevent accidental changes to set points
Smart2Pure Pro offers reservoir options, either 30 or 60L:
  • Large opening allows easy and effective cleaning by hand.
  • Sterile venting filter/reservoir overflow protects against contamination by microorganisms.
  • Optional CO2 absorber prevents TOC value increases from drawn-in CO2.
  • Polyethylene reservoir is opaque to light.
  • Recirculation pump protects the high purity water from bacterial growth during standstills and maintains the low conductivity value.
  • Wall-mount is optional for simple space-savings.
  • Conical bottom outlet allows for complete draining and efficient cleaning and disinfection. 
Optional Hand Dispenser:
  • Gain increased flexibility by adding a hand dispenser with 3m cord (Part #50138221)
Warranty: 1 year, parts and labor
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.99
<0.01 CFU/mL
No
Up to 1 L/min.
No
Water Purification System
Complete system includes Barnstead Pretreatment system with cartridges, RO/pretreatment cartridge, an ultrapure polisher cartridge, sterile 0.1 μm filter, UV lamp, UF filter, 30 L tank, CO2 vent filter, sterile overflow and system wall bracket
1 to 6 bar
16 L/hr. at 10°C
16 L/hr.
545 mm
22 kg
100/240 V
Type 1: 0.055μS/cm and Type II: 0.067 to 0.1μS/cm
3/4 in. NPT and 1/2 in. NPT
Tap
Type 1 and 2
1 to 5 ppb
1 to 6 bar
Smart2Pure Pro 16 UV/UF with 30L Tank - Complete system
<1
0.12 kW
305 mm
300 mm
100/240 V 50/60 Hz
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products