Axygen™ MultiRack Reload Tip System
MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in Bulk, Racked, Filter and Reload packaging styles to meet the variety of needs in the lab setting.
Additional Details:
Weight:
0.60300kg
Specification
Volume (Metric) | 300 μL |
---|---|
Filtered | Non-filtered |
Sterility | Non-sterile |
Color | Green |
Rack Color | Green |
Autoclavable | Autoclavable |
Certifications/Compliance | ISO 13845 |
Clean Claims | Free of detectable DNAse/RNAse, human DNA, and PCR inhibition |
Format | Refill |
Tip Style | Universal |
Packaging | Reload System |
Quantity | 960/pk, 4 pks/cs |
Disposable | Single-use |
Graduated | Yes |
Product Line | MultiRack |
Material | Polyethylene |