Merck Millex™-HV Sterile Syringe Filter Unit, PVDF, 0.45 μm (Research Use Only)
Ready-to-use, syringe-driven units designed for sterilizing and clarifying sterile solutions are available in 0.1, 0.22, 0.45 and 5.0 μm pore sizes.
Additional Details:
Weight:
0.36700kg
Specification
Autoclavable | Not Autoclavable |
---|---|
Color | Yellow |
Disposable | Single-use |
Material (Membrane) | PVDF (Hydrophilic) |
Pore Size | 0.45 μm |
Sterility | Sterile |
For Use With (Application) | Sterilization of tissue culture media, protein solutions or aqueous solutions;Low Protein Binding |
Hold up Volume (After Air Purge) | <100 μL |
Housing Material | PVDF |
Inlet Style | Female Luer Lock |
Outlet Style | Male Luer Slip |
Pressure Rating | 145 psi |
Quantity | 50/Pk. |
Temperature (English) Operating | 113°F |
Temperature (Metric) Operating | 45°C |
Temperature Range (English) | 113°F |
Temperature Range (Metric) | 45°C |
Tip Material | Modified Acrylic |
Type | Syringe Filter |
Volume (Metric) | 100 mL |
Diameter (Metric) | 3.3 cm |