Use promo code "24112" to get your promotional price.
Invitrogen™ E-Gel™ EX Agarose Gels
Invitrogen E-Gel EX precast agarose gels are designed for ultrasensitive and convenient DNA and RNA sample electrophoresis in as little as 10 to 20 minutes.
220.67 EUR valid until 2025-06-21
Use promo code "24112" to get your promotional price.
Use promo code "24112" to get your promotional price.
Additional Details:
Weight:
0.20000kg
Requires:
Requires the E-Gel iBase™ Power System for running and the E-Gel Safe Imager™ Real-Time Transilluminator for real-time viewing of sample migration.
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specification
Compatible Ladders | E-Gel™ 1 Kb Plus Express Ladder, Century™-Plus RNA Markers |
---|---|
Content And Storage |
• E-Gel™ EX 20-Paks contain 20 gels Store gels at room temperature. Guaranteed stable for 3 months when properly stored. |
Gel Percentage | 2% |
Gel Type | E-Gel |
Product Type | EX Agarose Gel |
Sample Loading Volume | 20 μL |
Separation Range | 50 to 2000 bp |
Stain Type | SYBR™ Gold |
Wells | 11-well |
Mode of Separation | Molecular Weight |
Quantity | 20 Gels/Pk. |
Sample Buffer Type | Preparative without dyes, Non-preparative |
Sample Type | Double-Stranded DNA (dsDNA), Single-Stranded RNA, Circular RNA |
Shipping Condition | Room Temperature |
Well Design | Single Comb |