Learn More
Velp Scientifica™ AREC 7 Digital Ceramic Hotplate Stirrer
AREC 7 Digital Ceramic Hotplate Stirrer is a safe and reliable Hotplate stirrer with ceramic top to guarantee excellent heat transfer and stirring performance. Excellent for basic laboratory applications.
Brand: Velp Scientifica™ F20500551
434.15 EUR valid until 2025-12-31
Use promo code "24301" to get your promotional price.
Description
Excellent Performance
- Heating plate technology ensures temperature homogeneity and stability from room temperature to 550°C
- Bright and easy-to-read digital display showing actual and setpoint temperature
- Powerful motor stirs up to 15 L volume (H2O) from 50 to 1500 rpm
Safe and Resistant
- Ceramic top and technopolymer structure ensure premium resistance to chemicals
- Hot Surface Safety message displayed when temperature is above 50°C
- Run-off groove to protect electronics
Specifications
Ceramic | |
Hot surface safety message | |
3.9 kg | |
800 W | |
7 x 7 in. | |
50/60 Hz | |
3.5 kg | |
Square | |
EU | |
15 L | |
25 Months | |
CE Declaration of Conformity, UK Declaration of Conformity, BC Certificate, cNEMKOus Certificate |
230 V | |
8.6 lb. | |
Technopolymer | |
Analog | |
Ambient to 550°C | |
7.7 lb. | |
Stirring Hotplate | |
25 kg | |
1 | |
50 to 1500 rpm | |
344 x 203 x 94 mm | |
Digital |